Gene Transcripts

Gene Symbol
Sh2b1
Entrez Gene ID
89817
Gene Name
SH2B adaptor protein 1
Gene Aliases
Sh2-b, Sh2b, Sh2bpsm1
Chromosome Location
Chr. 1: 197878839 - 197888215 on Build Rnor_6.0
UniGene ID
Rn.11069
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048180.1 NP_001041645.1 1 741
NM_134456.3 NP_604451.2 2 1041
XM_006230351.2 XP_006230413.1 1 2602
XM_006230352.2 XP_006230414.1 1 2602
XM_006230353.2 XP_006230415.1 2 1016
XM_006230354.2 XP_006230416.1 2 1922
XM_006230355.2 XP_006230417.1 1 2601
GenBank mRNA AF047577.1 - 1 741
U57391.1 - 2 1084

View Full Product Details