Gene Transcripts

Gene Symbol
Osgin1
Entrez Gene ID
171493
Gene Name
oxidative stress induced growth inhibitor 1
Gene Aliases
Okl38
Chromosome Location
Chr. 19: 52056816 - 52085491 on Build Rnor_6.0
UniGene ID
Rn.15599
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138504.2 NP_612513.1 8 1082
XM_006255706.2 XP_006255768.1 11 1141
XM_006255707.2 XP_006255769.1 7 1177
XM_008772594.1 XP_008770816.1 6 1003
GenBank mRNA AF549441.1 - 8 1082
AF549442.1 - 7 983
AY081218.1 - 6 731
BC091111.1 - 8 1078 7106386

View Full Product Details