Gene Transcripts

Gene Symbol
Slc38a1
Entrez Gene ID
170567
Gene Name
solute carrier family 38, member 1
Gene Aliases
Ata1, GlnT, Sat1
Chromosome Location
Chr. 7: 137970555 - 138040103 on Build Rnor_6.0
UniGene ID
Rn.162022
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138832.1 NP_620187.1 3, 4 423
XM_006242277.2 XP_006242339.1 3, 4 482
XM_006242278.2 XP_006242340.1 4, 5 691
XM_006242279.2 XP_006242341.1 4, 5 661
XM_006242280.2 XP_006242342.1 3, 4 530
XM_006242281.2 XP_006242343.1 3, 4 571
GenBank mRNA AF075704.1 - 3, 4 423
BC097283.1 - 3, 4 343 7444889

View Full Product Details