Gene Transcripts

Gene Symbol
Tgoln2
Entrez Gene ID
192152
Gene Name
trans-golgi network protein 2
Gene Aliases
Tgn38, Tgoln1, Ttgn1
Chromosome Location
Chr. 4: 100475415 - 100483168 on Build Rnor_6.0
UniGene ID
Rn.11349
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138840.2 NP_620195.1 2 200
GenBank mRNA BC098512.1 - 2 200
X53565.1 - 2 198
X64600.1 - 2 198

View Full Product Details