Gene Transcripts

Gene Symbol
Unc50
Entrez Gene ID
192356
Gene Name
unc-50 homolog (C. elegans)
Gene Aliases
Uncl
Chromosome Location
Chr. 9: 44024972 - 44032896 on Build Rnor_6.0
UniGene ID
Rn.3446
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138919.2 NP_620274.1 2 427
XM_006244736.2 XP_006244798.1 2 380
XM_006244737.2 XP_006244799.1 2 325
GenBank mRNA AY017209.1 - 2 427
BC061976.1 - 2 422 5623807
FQ220352.1 - 2 321
U96638.1 - 1 193

View Full Product Details