Gene Transcripts

Gene Symbol
Ppap2c
Entrez Gene ID
246115
Gene Name
phosphatidic acid phosphatase type 2c
Gene Aliases
Chromosome Location
Chr. 7: 13062178 - 13070281 on Build Rnor_6.0
UniGene ID
Rn.3301
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139252.1 NP_640345.1 3 390
XM_008765155.1 XP_008763377.1 3 373
GenBank mRNA AF503611.1 - 3 390
BC062088.1 - 3 346 5621318

View Full Product Details