Gene Transcripts

Gene Symbol
Baalc
Entrez Gene ID
140720
Gene Name
brain and acute leukemia, cytoplasmic
Gene Aliases
Chromosome Location
Chr. 7: 77763246 - 77836534 on Build Rnor_6.0
UniGene ID
Rn.19969
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144762.2 NP_658907.1 2, 3 629
GenBank mRNA AB073318.1 - 2, 3 629
AF371321.1 - 2, 3 335

View Full Product Details