Gene Transcripts

Gene Symbol
Olfm3
Entrez Gene ID
252920
Gene Name
olfactomedin 3
Gene Aliases
Chromosome Location
Chr. 2: 217963376 - 218005244 on Build Rnor_6.0
UniGene ID
Rn.27711
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145777.1 NP_665720.1 6 1338
GenBank mRNA AF442822.1 - 6 1338
AF442823.1 - 1229

View Full Product Details