Gene Transcripts

Gene Symbol
Tor1a
Entrez Gene ID
266606
Gene Name
torsin family 1, member A
Gene Aliases
Dyt1
Chromosome Location
Chr. 3: 9800322 - 9807318 on Build Rnor_6.0
UniGene ID
Rn.20041
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153303.2 NP_695215.2 2 238
GenBank mRNA AY054303.1 - 2 220
BC078714.1 - 2 238 7134604

View Full Product Details