Gene Transcripts
- Gene Symbol
- Prl3a1
- Entrez Gene ID
- 266782
- Gene Name
- Prolactin family 3, subfamily a, member 1
- Gene Aliases
- PLP-I, Plpi
- Chromosome Location
- Chr. 17: 42723745 - 42740021 on Build Rnor_6.0
- UniGene ID
- Rn.22898
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153736.1 |
NP_714958.1
|
2, 3 |
427 |
|
GenBank mRNA |
AF526270.1 |
- |
2, 3 |
427 |
|
View Full Product Details