Gene Transcripts

Gene Symbol
Prop1
Entrez Gene ID
266738
Gene Name
PROP paired-like homeobox 1
Gene Aliases
Chromosome Location
Chr. 10: 36449920 - 36452381 on Build Rnor_6.0
UniGene ID
Rn.86655
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153627.1 NP_705891.1 2 271
XM_008767664.1 XP_008765886.1 2 268
GenBank mRNA AB037922.1 - 2 271

View Full Product Details