Gene Transcripts

Gene Symbol
Prl3a1
Entrez Gene ID
266782
Gene Name
Prolactin family 3, subfamily a, member 1
Gene Aliases
PLP-I, Plpi
Chromosome Location
Chr. 17: 42723745 - 42740021 on Build Rnor_6.0
UniGene ID
Rn.22898
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153736.1 NP_714958.1 4 667
GenBank mRNA AB019791.1 - 4 669
AF526270.1 - 4 667

View Full Product Details