Gene Transcripts
- Gene Symbol
- Tpbpa
- Entrez Gene ID
- 64509
- Gene Name
- trophoblast specific protein alpha
- Gene Aliases
- Tpbp
- Chromosome Location
- Chr. 17: 4165041 - 4167411 on Build Rnor_6.0
- UniGene ID
- Rn.8816
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172073.1 |
NP_742070.1
|
2, 3 |
182 |
|
GenBank mRNA |
AB009890.1 |
- |
2, 3 |
182 |
|
View Full Product Details