Gene Transcripts
- Gene Symbol
- Hsbp1
- Entrez Gene ID
- 286899
- Gene Name
- heat shock factor binding protein 1
- Gene Aliases
- Hsp25
- Chromosome Location
- Chr. 19: 51985196 - 51989528 on Build Rnor_6.0
- UniGene ID
- Rn.2578
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173119.2 |
NP_775142.1
|
2, 3 |
206 |
|
GenBank mRNA |
AF522937.1 |
- |
2, 3 |
153 |
|
BC058131.1 |
- |
2, 3 |
191 |
6919748 |
EX491168.1 |
- |
2, 3 |
206 |
|
FQ211891.1 |
- |
2, 3 |
174 |
|
FQ212621.1 |
- |
2, 3 |
178 |
|
FQ213728.1 |
- |
2, 3 |
148 |
|
FQ216175.1 |
- |
2, 3 |
185 |
|
FQ229155.1 |
- |
2, 3 |
191 |
|
FQ229754.1 |
- |
2, 3 |
165 |
|
FQ230047.1 |
- |
2, 3 |
163 |
|
FQ230722.1 |
- |
2, 3 |
165 |
|
FQ233773.1 |
- |
2, 3 |
164 |
|
View Full Product Details