Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145846.2 | NP_001139318.1 | 4 | 306 | |
NM_172077.2 | NP_742074.2 | 4 | 408 | ||
GenBank mRNA | CF249035.1 | - | 3 | 295 | |
D23676.1 | - | 4 | 306 | ||
L10229.1 | - | 4 | 408 |