Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020988.2 | NP_066268.1 | 2 | 1038 | |
NM_138736.2 | NP_620073.2 | 2 | 1038 | ||
XM_011523003.2 | XP_011521305.1 | 1 | 24 | ||
GenBank mRNA | AF493894.1 | - | 2 | 141 | |
AF493895.1 | - | 2 | 141 | ||
AK056008.1 | - | 2 | 987 | ||
BC030027.2 | - | 2 | 74 | 4791738 | |
CR936770.1 | - | 2 | 401 | ||
KR081949.1 | - | 2 | 141 |