Gene Transcripts

Gene Symbol
Cxcl1
Entrez Gene ID
81503
Gene Name
chemokine (C-X-C motif) ligand 1
Gene Aliases
CINC-1, Gro1
Chromosome Location
Chr. 14: 18743678 - 18745457 on Build Rnor_6.0
UniGene ID
Rn.10907
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030845.1 NP_110472.1 2 174
GenBank mRNA D11444.1 - 2 159
M86536.1 - 2 174

View Full Product Details