Gene Transcripts
- Gene Symbol
- Tmem14c
- Entrez Gene ID
- 171432
- Gene Name
- transmembrane protein 14C
- Gene Aliases
- Cdtw1
- Chromosome Location
- Chr. 17: 21571510 - 21577652 on Build Rnor_6.0
- UniGene ID
- Rn.114642
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF083396.1 |
- |
|
669 |
|
View Full Product Details