Gene Transcripts

Gene Symbol
Foxa2
Entrez Gene ID
25099
Gene Name
forkhead box A2
Gene Aliases
Hnf3b
Chromosome Location
Chr. 3: 142383093 - 142387493 on Build Rnor_6.0
UniGene ID
Rn.10948
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012743.1 NP_036875.1 3 745
XM_006235139.2 XP_006235201.1 2 620
XM_006235140.2 XP_006235202.1 2 872
GenBank mRNA L09647.1 - 3 745

View Full Product Details