Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013090.2 | NP_037222.2 | 2 | 231 | |
GenBank mRNA | AF498262.1 | - | 1 | 97 | |
BC092206.1 | - | 2 | 129 | 7313343 | |
CB582258.1 | - | 2 | 231 | ||
CO567426.1 | - | 260 | |||
EF653274.1 | - | 2 | 156 | ||
EF653275.1 | - | 2 | 156 | ||
M24104.1 | - | 2 | 194 | ||
U74621.1 | - | 2 | 129 |