Gene Transcripts

Gene Symbol
Atp5g3
Entrez Gene ID
114630
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex, subunit C3 (subunit 9)
Gene Aliases
Chromosome Location
Chr. 3: 60811218 - 60813899 on Build Rnor_6.0
UniGene ID
Rn.2180
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053756.1 NP_446208.1 2 215
GenBank mRNA AF315374.1 - 2 215

View Full Product Details