Gene Transcripts

Gene Symbol
Adh1
Entrez Gene ID
11522
Gene Name
alcohol dehydrogenase 1 (class I)
Gene Aliases
ADH-AA, AI194826, Adh-1, Adh-1-t, Adh-1e, Adh-1t, Adh-3e, Adh1-e, Adh1-t, Adh1tl, Adh3-e
Chromosome Location
Chr. 3: 138277645 - 138290691 on Build GRCm38
UniGene ID
Mm.2409
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007409.2 NP_031435.1 2 181
GenBank mRNA AK146094.1 - 2 182
BC013477.1 - 2 181 4238555
BC054467.1 - 2 161 6530906

View Full Product Details