Gene Transcripts
- Gene Symbol
- Arf2
- Entrez Gene ID
- 11841
- Gene Name
- ADP-ribosylation factor 2
- Gene Aliases
- Chromosome Location
- Chr. 11: 103966718 - 103985337 on Build GRCm38
- UniGene ID
- Mm.5061
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK031259.1 |
- |
3, 4 |
337 |
|
AK042977.1 |
- |
3, 4 |
377 |
|
AK053123.1 |
- |
3, 4 |
795 |
|
AK077591.1 |
- |
3, 4 |
350 |
|
AK167133.1 |
- |
3, 4 |
347 |
|
AK167601.1 |
- |
3, 4 |
351 |
|
AK167653.1 |
- |
3, 4 |
350 |
|
AK169094.1 |
- |
3, 4 |
347 |
|
AK169412.1 |
- |
3, 4 |
347 |
|
BC010487.1 |
- |
3, 4 |
630 |
3483748 |
D87899.1 |
- |
3, 4 |
342 |
|
View Full Product Details