Gene Transcripts

Gene Symbol
Atp6v1e1
Entrez Gene ID
11973
Gene Name
ATPase, H+ transporting, lysosomal V1 subunit E1
Gene Aliases
2410029D23Rik, Atp6e, Atp6e2, Atp6v1e, D6Ertd385e, E2, P31, Vma4
Chromosome Location
Chr. 6: 120795244 - 120822685 on Build GRCm38
UniGene ID
Mm.29045
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007510.2 NP_031536.2 2, 3 172
GenBank mRNA AB074758.1 - 2, 3 174
AK010603.1 - 2, 3 189
AK139791.1 - 2, 3 189
AK146162.1 - 2, 3 186
AK149356.1 - 2, 3 208
AK167644.1 - 2, 3 131
AK169854.1 - 2, 3 186
AK186366.1 - 2, 3 124
AK189499.1 - 2, 3 121
BC003421.1 - 2, 3 172 2598896
BC055438.1 - 2, 3 206 3995744

View Full Product Details