Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301424.1 | NP_001288353.1 | 5 | 724 | |
NM_007513.4 | NP_031539.3 | 5 | 840 | ||
GenBank mRNA | AK139342.1 | - | 5 | 816 | |
AK147991.1 | - | 5 | 749 | ||
AK163202.1 | - | 5 | 724 | ||
BC068234.1 | - | 5 | 732 | ||
BC145779.1 | - | 4 | 645 | 40131082 | |
BC145781.1 | - | 4 | 645 | 40131084 | |
JN226407.1 | - | 3 | 560 | ||
JN226408.1 | - | 3 | 560 | ||
JN226410.1 | - | 3 | 560 | ||
M26687.1 | - | 5 | 758 |