Gene Transcripts

Gene Symbol
Psg17
Entrez Gene ID
26437
Gene Name
pregnancy specific glycoprotein 17
Gene Aliases
Cea-2, Cea2, mmCGM5
Chromosome Location
Chr. 7: 18813937 - 18821591 on Build GRCm38
UniGene ID
Mm.302844
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007677.2 NP_031703.1 3 752
GenBank mRNA BC100345.1 - 3 749
M83344.1 - 3 768

View Full Product Details