Gene Transcripts

Gene Symbol
Ccr1l1
Entrez Gene ID
12770
Gene Name
chemokine (C-C motif) receptor 1-like 1
Gene Aliases
Cmkbr1l1
Chromosome Location
Chr. 9: 123976949 - 123978408 on Build GRCm38
UniGene ID
Mm.423532
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007718.3 NP_031744.3 1 100
GenBank mRNA BC127143.1 - 1 100
BC128466.1 - 1 100 40051965

View Full Product Details