Gene Transcripts

Gene Symbol
Drd3
Entrez Gene ID
13490
Gene Name
dopamine receptor D3
Gene Aliases
D3R
Chromosome Location
Chr. 16: 43754015 - 43823846 on Build GRCm38
UniGene ID
Mm.439735
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007877.1 NP_031903.1 2 285
GenBank mRNA BC119286.1 - 2 306
BC137819.1 - 2 306 8860838
X67274.1 - 2 285

View Full Product Details