Gene Transcripts

Gene Symbol
NME4
Entrez Gene ID
4833
Gene Name
NME/NM23 nucleoside diphosphate kinase 4
Gene Aliases
NDPK-D, NM23H4, nm23-H4
Chromosome Location
Chr. 16: 396755 - 400754 on Build GRCh38
UniGene ID
Hs.9235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286433.1 NP_001273362.1 4 538
NM_001286435.1 NP_001273364.1 5 750
NM_001286436.1 NP_001273365.1 5 843
NM_001286438.1 NP_001273367.1 6 818
NM_001286439.1 NP_001273368.1 6 827
NM_001286440.1 NP_001273369.1 5 956
NM_005009.2 NP_005000.1 5 640
GenBank mRNA AF086133.1 - 3 338
AK094439.1 - 2 1499
BC004880.2 - 5 623 3830205
BC017067.1 - 5 640 3913604
Y07604.1 - 5 620

View Full Product Details