Gene Transcripts
- Gene Symbol
- Eps8
- Entrez Gene ID
- 13860
- Gene Name
- epidermal growth factor receptor pathway substrate 8
- Gene Aliases
- AW261790
- Chromosome Location
- Chr. 6: 137477245 - 137649397 on Build GRCm38
- UniGene ID
- Mm.235346
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271587.1 |
NP_001258516.1
|
3, 4 |
458 |
|
NM_001271588.1 |
NP_001258517.1
|
3, 4 |
389 |
|
NM_001271595.1 |
NP_001258524.1
|
3, 4 |
463 |
|
NM_007945.3 |
NP_031971.2
|
4, 5 |
464 |
|
GenBank mRNA |
AK147877.1 |
- |
4, 5 |
392 |
|
AK148424.1 |
- |
4, 5 |
380 |
|
AK149683.1 |
- |
3, 4 |
252 |
|
AK155022.1 |
- |
3, 4 |
224 |
|
AK166156.1 |
- |
4, 5 |
299 |
|
BC016890.1 |
- |
3, 4 |
475 |
4240899 |
BY209758.1 |
- |
3, 4 |
224 |
|
L21671.1 |
- |
3, 4 |
379 |
|
View Full Product Details