Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146348.1 | NP_001139820.1 | 11 | 1685 | |
NM_001146350.1 | NP_001139822.1 | 11 | 1688 | ||
NM_007932.2 | NP_031958.2 | 11 | 1688 | ||
GenBank mRNA | AK151302.1 | - | 11 | 1658 | |
AY679531.1 | - | 11 | 1688 | ||
BC029080.1 | - | 11 | 1602 | 4168250 | |
S69407.1 | - | 12 | 1553 | ||
X77952.1 | - | 11 | 1688 |