Gene Transcripts
- Gene Symbol
- Fpr3
- Entrez Gene ID
- 14294
- Gene Name
- formyl peptide receptor 3
- Gene Aliases
- ALX, Fpr-rs1, Fpr-s1, Fprl1, LXA4-R, Lxa4r
- Chromosome Location
- Chr. 17: 17970458 - 17971677 on Build GRCm38
- UniGene ID
- Mm.378918
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008042.2 |
NP_032068.2
|
1 |
438 |
|
GenBank mRNA |
U78299.1 |
- |
|
486 |
|
View Full Product Details