Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008123.3 | NP_032149.1 | 2 | 1358 | |
GenBank mRNA | BC120513.1 | - | 2 | 1279 | |
BC125569.1 | - | 2 | 1279 | 40130084 | |
GU363513.1 | - | 2 | 1237 | ||
GU363514.1 | - | 2 | 1327 | ||
GU363515.1 | - | 1 | 1544 |