Gene Transcripts
- Gene Symbol
- Grin2a
- Entrez Gene ID
- 14811
- Gene Name
- glutamate receptor, ionotropic, NMDA2A (epsilon 1)
- Gene Aliases
- GluN2A, NMDAR2A, NR2A
- Chromosome Location
- Chr. 16: 9573737 - 9996500 on Build GRCm38
- UniGene ID
- Mm.2953
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008170.2 |
NP_032196.2
|
1, 2 |
422 |
|
GenBank mRNA |
D10217.2 |
- |
1, 2 |
422 |
|
View Full Product Details