Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277970.1 | NP_001264899.1 | 18 | 2572 | |
NM_008397.4 | NP_032423.2 | 18 | 2572 | ||
GenBank mRNA | AK034186.1 | - | 18 | 2429 | |
AK045391.1 | - | 11 | 1153 | ||
AK159653.1 | - | 13 | 1508 | ||
AK193318.1 | - | 4 | 240 | ||
BC049132.1 | - | 19 | 2497 | ||
BC058095.1 | - | 19 | 2497 | 6826609 | |
X69902.1 | - | 18 | 2553 |