Gene Transcripts

Gene Symbol
VN1R10P
Entrez Gene ID
387316
Gene Name
vomeronasal 1 receptor 10 pseudogene
Gene Aliases
FKSG83, VNR6I1P, b24o18.2, hs6V1-1p
Chromosome Location
Chr. 6: 27324761 - 27325963 on Build GRCh38
UniGene ID
Hs.326753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_045612.1 1 520
GenBank mRNA AF336874.1 - 1 481
AF352327.1 - 1 499
BC107050.2 - 1 482 40004452

View Full Product Details