Gene Transcripts
- Gene Symbol
- Mme
- Entrez Gene ID
- 17380
- Gene Name
- membrane metallo endopeptidase
- Gene Aliases
- 6030454K05Rik, C85356, CALLA, CD10, NEP, SFE
- Chromosome Location
- Chr. 3: 63295226 - 63383713 on Build GRCm38
- UniGene ID
- Mm.296022
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001289462.1 |
NP_001276391.1
|
8, 9 |
1182 |
|
NM_001289463.1 |
NP_001276392.1
|
8, 9 |
966 |
|
NM_008604.4 |
NP_032630.2
|
8, 9 |
816 |
|
GenBank mRNA |
AK031446.1 |
- |
8, 9 |
817 |
|
AK033824.1 |
- |
8, 9 |
877 |
|
AK040730.1 |
- |
8, 9 |
853 |
|
AK052088.1 |
- |
7, 8 |
703 |
|
AK076908.1 |
- |
8, 9 |
1030 |
|
AK080202.1 |
- |
8, 9 |
844 |
|
BC034092.1 |
- |
8, 9 |
838 |
4483558 |
BC066840.1 |
- |
7, 8 |
725 |
|
M81591.1 |
- |
7, 8 |
728 |
|
View Full Product Details