Gene Transcripts
- Gene Symbol
- Nkx2-9
- Entrez Gene ID
- 18094
- Gene Name
- NK2 homeobox 9
- Gene Aliases
- Nkx-2.9, Nkx2-8, Nkx2.9, tinman
- Chromosome Location
- Chr. 12: 56611389 - 56614676 on Build GRCm38
- UniGene ID
- Mm.129420
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008701.2 |
NP_032727.2
|
2 |
1145 |
|
GenBank mRNA |
Y15741.1 |
- |
3 |
1114 |
|
View Full Product Details