Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008732.2 | NP_032758.2 | 17 | 1945 | |
GenBank mRNA | AK188425.1 | - | 1 | 167 | |
AK212997.1 | - | 1 | 167 | ||
AK215979.1 | - | 1 | 167 | ||
BC019137.1 | - | 17 | 1919 | 5040976 | |
L33415.1 | - | 17 | 1873 |