Gene Transcripts

Gene Symbol
Pdx1
Entrez Gene ID
18609
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
IDX-1, IPF-1, Ipf1, Mody4, STF-1, pdx-1
Chromosome Location
Chr. 5: 147270131 - 147275149 on Build GRCm38
UniGene ID
Mm.389714
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008814.3 NP_032840.1 2 731
GenBank mRNA AK020261.1 - 736
BC103572.1 - 2 731 40045668
BC103581.1 - 2 731 40045664
BC103582.1 - 2 731 40045666
BC105642.1 - 2 731 40045665
X74342.1 - 2 902

View Full Product Details