Gene Transcripts

Gene Symbol
Pet2
Entrez Gene ID
18630
Gene Name
plasmacytoma expressed transcript 2
Gene Aliases
PC231, PC326, Pex3
Chromosome Location
Chr. X: 89403848 - 89409689 on Build GRCm38
UniGene ID
Mm.3086
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008821.2 NP_032847.2 12 1825
GenBank mRNA BC138080.1 - 11 1646
M95564.1 - 12 1830

View Full Product Details