Gene Transcripts

Gene Symbol
OR51E2
Entrez Gene ID
81285
Gene Name
olfactory receptor family 51 subfamily E member 2
Gene Aliases
OR51E3P, OR52A2, PSGR
Chromosome Location
Chr. 11: 4680171 - 4697846 on Build GRCh38
UniGene ID
Hs.501758
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030774.3 NP_110401.1 2 2265
GenBank mRNA AY033942.1 - 2 2261
BC020768.1 - 4 1703 4249775

View Full Product Details