Gene Transcripts
- Gene Symbol
- Pnn
- Entrez Gene ID
- 18949
- Gene Name
- pinin
- Gene Aliases
- AU045199, D12Ertd512e
- Chromosome Location
- Chr. 12: 59066919 - 59074017 on Build GRCm38
- UniGene ID
- Mm.22347
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
Y08701.1 |
- |
2 |
150 |
|
View Full Product Details