Gene Transcripts

Gene Symbol
Abcd3
Entrez Gene ID
19299
Gene Name
ATP-binding cassette, sub-family D (ALD), member 3
Gene Aliases
AI313901, AU018866, AW146054, PMP68, PMP70, Pxmp1
Chromosome Location
Chr. 3: 121758907 - 121815215 on Build GRCm38
UniGene ID
Mm.399042
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008991.2 NP_033017.2 3 259
GenBank mRNA AK004660.1 - 3 240
AK144060.1 - 3 241
AK153298.1 - 3 217
BC009119.1 - 3 251 3154529
BC054446.1 - 3 207 6477711
BY123916.1 - 3 259

View Full Product Details