Gene Transcripts

Gene Symbol
Rab1
Entrez Gene ID
19324
Gene Name
RAB1, member RAS oncogene family
Gene Aliases
Gtbp, Rab-1, Rab1A, Ypt1, mKIAA3012
Chromosome Location
Chr. 11: 20201602 - 20226856 on Build GRCm38
UniGene ID
Mm.271944
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008996.3 NP_033022.1 6 2071
GenBank mRNA AF226873.1 - 6 2071
AK034408.1 - 6 2239
AK129477.1 - 6 2231
AK152978.1 - 6 2242
AK159425.1 - 6 2238
AK166249.1 - 6 2248

View Full Product Details