Gene Transcripts

Gene Symbol
Rpl12
Entrez Gene ID
269261
Gene Name
ribosomal protein L12
Gene Aliases
E430018F03
Chromosome Location
Chr. 2: 32961712 - 32964045 on Build GRCm38
UniGene ID
Mm.250030
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009076.3 NP_033102.2 3 261
GenBank mRNA AK002973.1 - 3 260
AK008347.1 - 3 260
AK012349.1 - 3 260
AK012428.1 - 3 258
AK088464.1 - 3 260
AK144905.1 - 3 260
AK148566.1 - 462
AK150318.1 - 3 261
AK151532.1 - 3 260
AK151592.1 - 3 260
AK167761.1 - 3 280
AK168121.1 - 3 260
AK194205.1 - 1 21
AK198056.1 - 1 21
BC018321.1 - 3 257 3583073
BC075731.1 - 2 449
BC081469.1 - 3 244 6824411
BY295120.1 - 3 263
L04280.1 - 3 248

View Full Product Details