Gene Transcripts

Gene Symbol
Rdh16
Entrez Gene ID
19683
Gene Name
retinol dehydrogenase 16
Gene Aliases
CRAD, CRAD1, Rdh6
Chromosome Location
Chr. 10: 127801090 - 127815839 on Build GRCm38
UniGene ID
Mm.100276
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009040.3 NP_033066.1 4 2601
GenBank mRNA AF030513.1 - 4 2602

View Full Product Details