Gene Transcripts

Gene Symbol
Resp18
Entrez Gene ID
19711
Gene Name
regulated endocrine-specific protein 18
Gene Aliases
AI851012
Chromosome Location
Chr. 1: 75272197 - 75278455 on Build GRCm38
UniGene ID
Mm.4032
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009049.1 NP_033075.1 4 438
GenBank mRNA AK012477.1 - 4 389
AK019440.1 - 4 388
BC027535.1 - 4 401 3376899
L34214.1 - 4 438

View Full Product Details