Gene Transcripts

Gene Symbol
Cxcl2
Entrez Gene ID
20310
Gene Name
chemokine (C-X-C motif) ligand 2
Gene Aliases
CINC-2a, GROb, Gro2, MIP-2, MIP-2a, Mgsa-b, Mip2, Scyb, Scyb2
Chromosome Location
Chr. 5: 90903899 - 90905938 on Build GRCm38
UniGene ID
Mm.4979
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009140.2 NP_033166.1 2 185
GenBank mRNA AK137628.1 - 2 213
AK150450.1 - 2 93
AK155458.1 - 1 304
AK155874.1 - 2 214
AK155916.1 - 2 212
AK157079.1 - 2 212
BC119511.1 - 2 159 40055368
BC119512.1 - 2 159 40055369
X53798.1 - 2 185

View Full Product Details