Gene Transcripts

Gene Symbol
St8sia2
Entrez Gene ID
20450
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2
Gene Aliases
AI323367, ST8SiaII, STX, Siat8b
Chromosome Location
Chr. 7: 73939119 - 74013727 on Build GRCm38
UniGene ID
Mm.4954
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009181.2 NP_033207.2 2 305
GenBank mRNA BC096546.1 - 2 297
X83562.1 - 2 284

View Full Product Details